Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
BDH7232-1
BDH7232-4
BDH7232-1CS
756.97
CAD
BDH7232-1
BDH7232-4
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Sulfuric acid
APHA. Made with high purity acid and deionized water. Suitable as a titrant.
Formula:
H₂SO₄ MW: 98.08 g/mol Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8,II |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. Traceable to SRM 723 | 1 |
Normality | 0.995 - 1.005 N |
Shelf-life | 2 years |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...