Sodium thiosulfate 0.10 N aqueous solution zA, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

BDH7228-4 BDH7228-7 BDH7228-1
BDH7228-4CS 620.87 CAD
BDH7228-4 BDH7228-7 BDH72281
Sodium thiosulfate 0.10 N aqueous solution zA, VWR Chemicals BDH®
Sodium thiosulphate

Suitable as a titrant. Made with deionized water.

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Density: 1.008 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 136 Confirmed
Normality 0.0995 - 0.1005 N
Prepared to formulation on file Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR