Sodium thiosulfate 0.025 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

BDH7227-1 BDH7227-4
BDH7227-1CS 1056.48 CAD
BDH7227-1 BDH7227-4
Sodium thiosulfate 0.025 N in aqueous solution, VWR Chemicals BDH®
Sodium thiosulphate

Suitable as a titrant. Made with deionized water.

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 136 Yes
Normality 0.0245 - 0.0255 N
Shelf-life 6 Months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR