Sodium hydroxide 5.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH7225-1
BDH7225-4
BDH7225-1CS
1138.09
CAD
BDH7225-1
BDH7225-4
Sodium hydroxide 5.0 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
Made with deionized water. Suitable as a titrant.
Formula:
NaOH Boiling Pt: 109 °C (1013 hPa) Density: ≤1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. traceable to SRM 84 | Yes |
Normality | 4.95 - 5.05 N |
Shelf-life | 2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...