Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
Caustic soda
BDH7222-1
BDH7222-4
BDH7222-7
BDH7222-1CS
871.89
CAD
BDH7222-1
BDH7222-4
BDH7222-7
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
NIST traceable. Suitable as a titrant.
Formula:
NaOH MW: 40 g/mol Density: 1.04 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 3266 ADR: 8,III |
Specification Test Results
Appearance/color | Clear liquid |
Traceable to NIST SRM 84 | X |
Normality | 0.995 - 1.005 N |
Shelf-life | 2 years |
Manufacture date | As reported |
Expiration date | As reported |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...