Sodium hydroxide 0.5 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Caustic soda

BDHVBDH7221-7 BDHVBDH7221-4 BDHVBDH7221-1
BDH7221-4CS 607.23 CAD
BDH7221-4 BDH7221-1 BDH7221-7
Sodium hydroxide 0.5 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

AOAC, APHA, and ASTM. Suitable as titrant.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Normality 0.499 - 0.501 N
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR