Sodium hydroxide ( 0.13 mol/l; 0.13 N) in aqueous solution 0.1 N aqueous solution zA, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Caustic soda

BDH7219-1 BDH7219-4 BDH7219-5 BDH7219-7
BDH7219-1CS 901.15 CAD
BDH7219-1 BDH7219-4 BDH7219-5 BDH7219-7
Sodium hydroxide ( 0.13 mol/l; 0.13 N) in aqueous solution 0.1 N aqueous solution zA, VWR Chemicals BDH®
Sodium hydroxide

AOAC, APHA, and ASTM. Suitable as titrant.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.003 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Normality 0.0995 - 0.1005 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR