Sodium hydroxide 20% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Caustic soda

BDHVBDH7216-4
BDH7216-4CS 763.05 CAD
BDH7216-4
Sodium hydroxide 20% (w/v) in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

AOAC and APHA.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Assay (w/v) 19.50 - 20.50 %
Shelf-life see CoA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR