Potassium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Potash causticCaustic potash

BDHVBDH7211-1
BDH7211-1CS 973.85 CAD
BDH7211-1
Potassium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Potassium hydroxide

Made with deionized water. Suitable as a titrant.

Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Normality 0.0095 - 0.1005 N
Shelf-life 18 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR