Hydrochloric acid 6.0 N, VWR Chemicals BDH®
Supplier: VWR International
|
Warning
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
BDH7204-1
BDH7204-4
BDH7204-1CS
2166.62
CAD
BDH7204-1
BDH7204-4
Hydrochloric acid 6.0 N, VWR Chemicals BDH®
Hydrochloric acid
Made with high purity acid and deionized water. Suitable as a titrant.
Formula:
HCl MW: 36.46 g/mol Density: 1.06 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Appearance/color | Clear liquid |
Normality | 5.950 - 6.050 N |
Shelf-life | 2 years |
N.I.S.T. traceable to SRM 723 | 1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...