Hydrochloric acid 2.0 N, VWR Chemicals BDH®
Supplier: VWR International
|
Warning
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
BDH7203-1
BDH7203-2
BDH7203-4
BDH7203-1CS
1193.62
CAD
BDH7203-1
BDH7203-2
BDH7203-4
Hydrochloric acid 2.0 N, VWR Chemicals BDH®
Hydrochloric acid
Made with high purity acid and deionized water. Suitable as a titrant.
Formula:
HCl MW: 36.46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. Traceable to SRM 723 | X |
Normality | 1.995 - 2.005 N |
Shelf-life | 2 years |
Certified total organic carbon (ppm) | 0.0 - 4 |
Colouration (APHA) | 10 max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...