Hydrochloric acid 1.0 N, VWR Chemicals BDH®
Supplier: VWR International
|
Warning
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
BDH7202-1
BDH7202-2
BDH7202-4
BDH7202-7
BDH7202-1CS
927.13
CAD
BDH7202-1
BDH7202-2
BDH7202-4
BDH7202-7
Hydrochloric acid 1.0 N, VWR Chemicals BDH®
Hydrochloric acid
Meets AOAC 936.15 standard and ASTM E200-91 standard. Made with high purity acid and deionized water. Suitable as a titrant.
Formula:
HCl MW: 36.46 Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8 II |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. Traceable to SRM 723 | Yes |
Normality | 0.995 - 1.005 N |
Shelf-life | 2 years |
Certified total organic carbon | 0.0 - 2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...