Hydrochloric acid 0.5 N, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

BDH7201-1 BDH7201-4 BDH7201-7
BDH7201-1CS 944.49 CAD
BDH7201-1 BDH7201-4 BDH7201-7
Hydrochloric acid 0.5 N, VWR Chemicals BDH®
Hydrochloric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 X
Normality 0.495 - .0505 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR