Hydrochloric acid (0.027 - 0.27 mol/l; 0.027 - 0.27 N) 0.10 N zA, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Hydrochloric acid solution, Muriatic acid

BDH7200-1 BDH7200-4 BDH7200-7
BDH7200-1CS 782.21 CAD
BDH7200-1 BDH7200-4 BDH7200-7
Hydrochloric acid (0.027 - 0.27 mol/l; 0.027 - 0.27 N) 0.10 N zA, VWR Chemicals BDH®
Hydrochloric acid

Meets AOAC 936.15 standard and ASTM E200-91 standard. Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
MW: 36.46
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 0.0995 - 0.1005 N
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR