Large Evaporator Flasks, Ace Glass Incorporated
Supplier: Ace Glass
6701-12 6701-22
89055-816EA 1396.63 CAD
89055-816 89055-818
Large Evaporator Flasks, Ace Glass Incorporated
Flasks Rotary Evaporator Flasks
These large evaporator flasks are used with Heidolph 20L rotary evaporators
They are fabricated from blanks selected for balance and quality. The necks are carefully welded to prevent “rotational whip.” Fabricated from borosilicate glass. Flange A O.D. is 143mm. Flange B O.D. is 118mm. Flange C O.D. is 18mm.