Whizard® Stainless Steel Mesh Glove with 7 ¹/₂" Cuff, Wells Lamont
Supplier: Wells Lamont
CM031901 CM031902 CM031903 CM031904 CM031905 CM031906
80091-076EA 990.82 CAD
80091-076 80091-078 80091-080 80091-082 80091-084 80091-086
Whizard® Stainless Steel Mesh Glove with 7 ¹/₂" Cuff, Wells Lamont
Gloves
Whizard® stainless steel gloves with 7¹/₂," cuff are ergonomically designed to be worn on both hands.
  • Ergonomically designed
  • Patented self-adjusting tensioning system
  • Micro plasma welded rings
  • No gaps to trap soil
  • Ambidextrous
  • Easy to clean
Whizard® stainless steel gloves provide superior cut and puncture protection. They feature a patented self-adjusting tensioning system on wrist allows for an easy fit with no straps or buckles to fasten. All Whizard® mesh gloves come with a metal serial number tag, which helps with reissuance and sizing information. Micro plasma welded rings have no gaps to trap soil that can feed bacterial growth. These gloves are made with 4 mm ring using 0.5 mm wire. Gloves are ambidextrous; just turn inside out for the other hand. These gloves are easy to clean.
Ordering information: These gloves are sold by the piece. Repair service is available to extend the life of the glove. For authorized repair center call 800-247-3295.