Dibenzyl azodicarboxylate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19347-06
L19347-14
CAAAAL19347-06EA
115.5
CAD
CAAAAL19347-06
CAAAAL19347-14
Dibenzyl azodicarboxylate 96%
Dibenzyl azodicarboxylate
Formula:
C₁₆H₁₄N₂O₄ Melting Pt: 43…47 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016737 CAS Number: 2449-05-0 EINECS: 219-508-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...