5-Bromo-3-indolyl-β-D-galactopyranoside ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Bluo-Gal, (2S,3R,4S,5R,6R)-2-((5-Bromo-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 5-Bromo-3-indolyl β-D-galactopyranoside

J65978-03 J65978-MC
CAAAJ65978-03EA 361.82 CAD
CAAAJ65978-03 CAAAJ65978-MC
5-Bromo-3-indolyl-β-D-galactopyranoside ≥98%
5-Bromo-3-indolyl-beta-D-galactopyranoside

alpha-Galactosidase substrate for Lac-gene detection systems


Powder


Applications: alpha-Galactosidase substrate for Lac-gene detection systems

Formula: C₁₄H₁₆BrNO₆
MW: 374.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063691
CAS Number: 97753-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR