2'-Deoxy-2'-fluorouridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
J65948-03
J65948-06
J65948-14
CAAAJ65948-03EA
55.3
CAD
CAAAJ65948-03
CAAAJ65948-06
CAAAJ65948-14
2'-Deoxy-2'-fluorouridine 97%
2'-Deoxy-2'-fluorouridine
Powder
Purity: 97%
CAS No.: 784-71-4
Molecular Formula: C9H11FN2O5
Formula Weight: 246.19
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD01317293
Formula:
C₉H₁₁FN₂O₅ MW: 246.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01317293 CAS Number: 784-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...