CPTH2

Supplier: Thermo Scientific Chemicals

J65939-LB0 J65939-MA
CAAAJ65939-LB0EA 138.88 CAD
CAAAJ65939-LB0 CAAAJ65939-MA
CPTH2
CPTH2

A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators


  • Light Sensitive


Solid


Soluble in DMSO

Formula: C₁₄H₁₄ClN₃S
Storage Temperature: Ambient
MDL Number: MFCD02307488
CAS Number: 357649-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR