Leflunomide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide

J65917-MC J65917-ME
CAAAJ65917-MCEA 249.64 CAD
CAAAJ65917-MC CAAAJ65917-ME
Leflunomide
Leflunomide

An immunosuppressive agent. Inhibits T and B cell proliferation.


Crystalline solid


Applications: An immunosuppressive agent. Inhibits T and B cell proliferation.

Formula: C₁₂H₉F₃N₂O₂
MW: 270.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00867593
CAS Number: 75706-12-6
UN: 2811
ADR: "6.1",III
Merck Index: 14,5432

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR