3'-Deoxythymidine (2',3'-dideoxythymidine)
Supplier: Thermo Scientific Chemicals
Synonyms:
3'-Deoxythymidine, 1-((2R,5S)-5-(Hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione
J65765-03
J65765-ME
CAAAJ65765-03EA
1166.04
CAD
CAAAJ65765-03
CAAAJ65765-ME
3'-Deoxythymidine (2',3'-dideoxythymidine)
3'-Deoxythymidine (2',3'-Dideoxythymidine)
Powder
CAS No.: 3416-05-5
Molecular Formula: C10H14N2O4
Formula Weight: 226.23
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 155-156°
Beilstein Reference: 21884
MDL No.: MFCD00010570
Formula:
C₁₀H₁₄N₂O₄ MW: 226.23 g/mol Melting Pt: 155…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010570 CAS Number: 3416-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...