Cytosine-β-D-arabinofuranoside hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Arabinosylcytosine hydrochloride, Cytarabine HCI, Cytarabine HCl, 4-Amino-1-((2S,3R,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride, 1-(β-D-Arabinofuranosyl)cytosine HCl

J65671-03 J65671-06 J65671-09
CAAAJ65671-03EA 92.11 CAD
CAAAJ65671-03 CAAAJ65671-06 CAAAJ65671-09
Cytosine-β-D-arabinofuranoside hydrochloride
Cytosine-beta-D-arabinofuranoside hydrochloride

Interferes with the synthesis of DNA


Powder


Applications: Interferes with the synthesis of DNA

Formula: C₉H₁₄ClN₃O₅
MW: 279.677 g/mol
Melting Pt: 197…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00012839
CAS Number: 69-74-9
EINECS: 200-713-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR