N-Acetyl-L-histidine monohydrate
Supplier: Thermo Scientific Chemicals
J65657-06
J65657-03
J65657-14
CAAAJ65657-06EA
327.06
CAD
CAAAJ65657-06
CAAAJ65657-03
CAAAJ65657-14
N-Acetyl-L-histidine monohydrate
N-Acetyl-L-histidine monohydrate
Powder
CAS No.: 39145-52-3
Molecular Formula: C8H11N3O3.H2O
Formula Weight: 215.21 (197.09anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 163°
Optical Rotation: +47°(c=1 in water)
MDL No.: MFCD00149320
Formula:
C₈H₁₁N₃O₃·H₂O Melting Pt: 163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149320 CAS Number: 39145-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...