2-Deoxy-D-galactose 99%
Supplier: Thermo Scientific Chemicals
J65482-04
J65482-06
J65482-09
CAAAJ65482-04EA
237
CAD
CAAAJ65482-04
CAAAJ65482-06
CAAAJ65482-09
2-Deoxy-D-galactose 99%
2-Deoxy-D-galactose
For studying galactose uptake into Escherichia coli
Powder
Applications: For studying galactose uptake into Escherichia coli
Formula:
C₆H₁₂O₅ Melting Pt: 107…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014649 CAS Number: 1949-89-9 EINECS: 217-765-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...