PYR-41

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-(4-((5-nitrofuran-2-yl)methylene)-3,5-dioxopyrazolidin-1-yl)benzoate

J65388-MB J65388-MC
CAAAJ65388-MBEA 350.76 CAD
CAAAJ65388-MB CAAAJ65388-MC
PYR-41
PYR-41

PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.


  • Light Sensitive


Solid


Soluble in DMSO at 5mg/ml with warming

Formula: C₁₇H₁₃N₃O₇·3H₂O
MW: 425.40 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01469983
CAS Number: 418805-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR