2'-Deoxycytidine-5'-monophosphate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
((2R,3S,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate, 2'-Deoxycytidine-5'-monophosphoric acid
J65244-03
J65244-06
CAAAJ65244-03EA
184.86
CAD
CAAAJ65244-03
CAAAJ65244-06
2'-Deoxycytidine-5'-monophosphate 99%
2'-Deoxycytidine-5'-monophosphate
Crystalline powder
Purity: 99%
CAS No.: 1032-65-1
Molecular Formula: C9H14N3O7P
Formula Weight: 307.20
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Appearance: White
Beilstein Reference: 41062
MDL No.: MFCD00006546
Formula:
C₉H₁₄N₃O₇P MW: 307.20 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00006546 CAS Number: 1032-65-1 EINECS: 213-849-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...