Bromocresol green ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
J65225-06
J65225-14
CAAAJ65225-06EA
69.38
CAD
CAAAJ65225-06
CAAAJ65225-14
Bromocresol green ACS
Bromocresol green
Powder
CAS No.: 76-60-8
Molecular Formula: C21H14Br4O5S
Formula Weight: 698.04
Physical Form: Powder
Appearance: Pale brown to yellow
Melting Point: 225° dec.
Beilstein Reference: 372527
Merck Reference: 14,1386
MDL No.: MFCD00005874
Formula:
C₂₁H₁₄Br₄O₅S MW: 698.02 g/mol Melting Pt: 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005874 CAS Number: 76-60-8 EINECS: 200-972-8 Merck Index: 14,1386 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...