2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium 2'-deoxy-5'-O-phosphonatoguanosine, 2'-Deoxyguanosine-5'-monophosphoric acid disodium salt

J65186-03 J65186-06 J65186-14
CAAAJ65186-03EA 109.81 CAD
CAAAJ65186-03 CAAAJ65186-06 CAAAJ65186-14
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate

Crystalline powder


Purity: 98% CAS No.: 33430-61-4 Molecular Formula: C10H12N5Na2O7P.xH2O Formula Weight: 391.18 (anhy) Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White Melting Point: 245° dec. MDL No.: MFCD00080331

Formula: C₁₀H₁₂N₅Na₂O₇P·xH₂O
MW: 391.18 g/mol (anhy)
Melting Pt: >245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00080331
CAS Number: 33430-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR