Cromolyn sodium 98%

Supplier: Thermo Scientific Chemicals
Warning

J65122-03 J65122-06 J65122-14
CAAAJ65122-03EA 93.54 CAD
CAAAJ65122-03 CAAAJ65122-06 CAAAJ65122-14
Cromolyn sodium 98%
Cromolyn sodium

Mast cell stabilizer that prevents release of histamine from mast cells


Powder


Applications: Mast cell stabilizer that prevents release of histamine from mast cells

Formula: C₂₃H₁₆O₁₁
Storage Temperature: Ambient
MDL Number: MFCD00057744
CAS Number: 15826-37-6
EINECS: 239-926-7
Merck Index: 14,2590

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR