N6-Methyl-2'-deoxyadenosine 99%
Supplier: Thermo Scientific Chemicals
J64961-03
J64961-MD
CAAAJ64961-03EA
856.36
CAD
CAAAJ64961-03
CAAAJ64961-MD
N6-Methyl-2'-deoxyadenosine 99%
N6-Methyl-2'-deoxyadenosine
Powder
Purity: 99%
CAS No.: 2002-35-9
Molecular Formula: C11H15N5O3
Formula Weight: 265.27
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water
MDL No.: MFCD00055999
Formula:
C₁₁H₁₅N₅O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00055999 CAS Number: 2002-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...