MIF Antagonist, ISO-1

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-(3-(4-hydroxyphenyl)-4,5-dihydroisoxazol-5-yl)acetate, ISO-1

J64947-LB0 J64947-MB
CAAAJ64947-LB0EA 176.96 CAD
CAAAJ64947-LB0 CAAAJ64947-MB
MIF Antagonist, ISO-1
MIF antagonist, ISO-1

A cell-permeable isoxazoline compound


Solid


A cell-permeable isoxazoline compound

Formula: C₁₂H₁₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD22376813
CAS Number: 478336-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR