2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64938-MB
J64938-MC
CAAAJ64938-MBEA
331.8
CAD
CAAAJ64938-MB
CAAAJ64938-MC
2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
2'-Deoxythymidine-5'-diphosphate trisodium salt
Powder
Purity: 97%
CAS No.: 95648-78-5
Molecular Formula: C10H13N2Na3O11P2
Formula Weight: 468.13
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00084748
Formula:
C₃₀H₄₅N₆Na₃O₃₃P₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00084748 CAS Number: 95648-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...