N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Supplier: Thermo Scientific Chemicals

Synonyms: Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, Bz-Arg-pNA·HCl, Bz-Arg-PNA.HCl, N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride, (S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

J64915-ME J64915-MC J64915-MD
CAAAJ64915-MEEA 458.2 CAD
CAAAJ64915-ME CAAAJ64915-MC CAAAJ64915-MD
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Chromogenic trypsin substrate. Substrate for proteolytic enzymes


Powder


Applications: Chromogenic trypsin substrate. Substrate for proteolytic enzymes

Formula: C₁₉H₂₃ClN₆O₄
MW: 434.881 g/mol
Melting Pt: 223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063682
CAS Number: 21653-40-7
EINECS: 244-505-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR