5,6-Dichlorobenzimidazole 1-beta-D-Ribofuranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5,6-Dichloro-1-(β-D-ribofuranosyl)benzimidazole, 5,6-Dichlorobenzimidazole Riboside, (2R,3R,4S,5R)-2-(5,6-Dichloro-1H-benzo[d]imidazol-1-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, DRB
J64870-MC
J64870-MD
J64870-MF
CAAAJ64870-MCEA
540.36
CAD
CAAAJ64870-MC
CAAAJ64870-MD
CAAAJ64870-MF
5,6-Dichlorobenzimidazole 1-beta-D-Ribofuranoside 98%
5,6-Dichlorobenzimidazole-1-beta-D-Ribofuranoside
Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.
Powder
Applications: Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.
Formula:
C₁₂H₁₂Cl₂N₂O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00036785 CAS Number: 53-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...