2',3'-Dideoxyinosine 98%
Supplier: Thermo Scientific Chemicals
J64833-03
J64833-MD
J64833-ME
CAAAJ64833-03EA
230.68
CAD
CAAAJ64833-03
CAAAJ64833-MD
CAAAJ64833-ME
2',3'-Dideoxyinosine 98%
2',3'-Dideoxyinosine
Powder
Purity: 98%
CAS No.: 69655-05-6
Molecular Formula: C10H12N4O3
Formula Weight: 236.23
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Solubility: Soluble in DMSO or methanol
Beilstein Reference: 3619529
Merck Reference: 14,3098
MDL No.: MFCD00077728
Formula:
C₁₀H₁₂N₄O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00077728 CAS Number: 69655-05-6 Merck Index: 14,3098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...