3-Methyladenine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methyl-3H-purin-6-amine, 3-Methyl-3H-adenine

J64813-MB J64813-MF
CAAAJ64813-MBEA 120.71 CAD
CAAAJ64813-MB CAAAJ64813-MF
3-Methyladenine
3-Methyladenine

A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation


  • Autophagy Inhibitor


White solid

Formula: C₆H₇N₅
MW: 149.16
Storage Temperature: Ambient
MDL Number: MFCD00010531
CAS Number: 5142-23-4
EINECS: 225-908-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR