Tripeptidyl Peptidase Inhibitor

Supplier: Thermo Scientific
J64760-LB0 J64760-LB0 J64760-MA J64760-MA
CAAAJ64760-MAEA 303.36 CAD
AAJ64760-MA CAAAJ64760-MA AAJ64760-LB0 CAAAJ64760-LB0
Tripeptidyl Peptidase Inhibitor
Proteins and Peptides
A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II

CAS No.: 184901-82-4
Molecular Formula: C16H22ClN3O3
Formula Weight: 453.90
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
MDL No.: MFCD01310850
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR