1,4-Dithioerythritol 99% for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64656-03
J64656-14
CAAAJ64656-03EA
75.84
CAD
CAAAJ64656-03
CAAAJ64656-14
1,4-Dithioerythritol 99% for electrophoresis
1,4-Dithioerythritol
Powder
Purity: 99%
CAS No.: 6892-68-8
Molecular Formula: C4H10O2S2
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: White
Odor: Stench
Melting Point: 82-84°
Beilstein Reference: 1719756
UN No.: 3335
MDL No.: MFCD00063750
Formula:
C₄H₁₀O₂S₂ Boiling Pt: 286 °C (1013 hPa) Melting Pt: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063750 CAS Number: 6892-68-8 EINECS: 229-998-8 UN: 3335 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...