2′-Deoxyuridine-5′-monophosphate disodium salt 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium ((2R,3S,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl phosphate, 2′-Deoxyuridine 5′-mono-phos-phate disodium salt
J64627-03
J64627-MD
CAAAJ64627-03EA
374.46
CAD
CAAAJ64627-03
CAAAJ64627-MD
2′-Deoxyuridine-5′-monophosphate disodium salt 99%
2′-Deoxyuridine-5′-monophosphate disodium salt
Powder
Purity: 99%
CAS No.: 42155-08-8
Molecular Formula: C9H11N2Na2O8P
Formula Weight: 352.15
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00065282
Formula:
C₉H₁₁N₂Na₂O₈P MW: 352.146 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00065282 CAS Number: 42155-08-8 EINECS: 255-687-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...