Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide)

Supplier: Thermo Scientific Chemicals
Warning

J64578-03 J64578-MD
CAAAJ64578-03EA 777.36 CAD
CAAAJ64578-03 CAAAJ64578-MD
Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide)
Deoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide)

Analog of CHAPS and CHAPSO.


Powder


Analog of CHAPS and CHAPSO

Formula: C₄₂H₇₅N₃O₁₅
Storage Temperature: Ambient
MDL Number: MFCD00161482
CAS Number: 86303-23-3
Merck Index: 14,1214

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR