Dithiothreitol (DTT, Cleland's reagent) 99% for electrophoresis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

J64545-03 J64545-06 J64545-14
CAAAJ64545-03EA 73.63 CAD
CAAAJ64545-03 CAAAJ64545-06 CAAAJ64545-14
Dithiothreitol (DTT, Cleland's reagent) 99% for electrophoresis
Dithiothreitol (DTT, Cleland's reagent)

Powder


Purity: 99% CAS No.: 3483-12-3 Molecular Formula: C4H10O2S2 Formula Weight: 154.25 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Sensitivity: Air Sensitive Appearance: White Odor: Unpleasant Melting Point: 40-43° Boiling Point: 125-130°/12mm Beilstein Reference: 1719757 Merck Reference: 14,3376 MDL No.: MFCD00004877

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 125…130 °C/12 mm
Melting Pt: 40…43 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
Merck Index: 14,3376
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR