Azure C
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Amino-7-(methylamino)phenothiazin-5-ium chloride
J64515-03
J64515-06
CAAAJ64515-03EA
30.49
CAD
CAAAJ64515-03
CAAAJ64515-06
Azure C
Azure C
Powder
Powder
Formula:
C₁₃H₁₂ClN₃S MW: 277.78 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00067677 CAS Number: 531-57-7 EINECS: 208-512-8 Merck Index: 14,929 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...