Adenosine-5'-diphosphate trilithium salt 98%

Supplier: Thermo Scientific Chemicals

J64497-03 J64497-06 J64497-14
CAAAJ64497-03EA 51.19 CAD
CAAAJ64497-03 CAAAJ64497-06 CAAAJ64497-14
Adenosine-5'-diphosphate trilithium salt 98%
Adenosine-5'-diphosphate trilithium salt

Powder


Purity: 98% CAS No.: 31008-64-7 Molecular Formula: C10H12Li3N5O10P2 Formula Weight: 445.00 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00065469

Formula: C₁₀H₁₂Li₃N₅O₁₀P₂
MW: 445.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065469
CAS Number: 31008-64-7
EINECS: 250-431-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR