Chloroquine diphosphate salt 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64459-22
J64459-14
J64459-18
J64459-30
CAAAJ64459-22EA
437.66
CAD
CAAAJ64459-22
CAAAJ64459-14
CAAAJ64459-18
CAAAJ64459-30
Chloroquine diphosphate salt 98%
Chloroquine diphosphate salt
A mild immunosupressant
Purity: 98%
CAS No.: 50-63-5
Molecular Formula: C18H26ClN3.2H3PO4
Formula Weight: 515.87
Physical Form: Powder
Appearance: White to off-white
Melting Point: 213-216°
Solubility: Soluble in water
Beilstein Reference: 4223142
MDL No.: MFCD00069852
Applications: A mild immunosupressant
Formula:
C₁₈H₂₆ClN₃ Melting Pt: 213…216 °C Storage Temperature: Ambient |
MDL Number:
MFCD00069852 CAS Number: 50-63-5 EINECS: 200-055-2 Merck Index: 14,2163 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...