L-Cysteine methyl ester hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64436-14
J64436-22
J64436-30
CAAAJ64436-14EA
100.49
CAD
CAAAJ64436-14
CAAAJ64436-22
CAAAJ64436-30
L-Cysteine methyl ester hydrochloride
L-Cysteine methyl ester hydrochloride
Crystalline powder
CAS No.: 18598-63-5
Molecular Formula: C4H9NO2S.HCl
Formula Weight: 171.65
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
Melting Point: 142° dec.
Solubility: Soluble in water
Beilstein Reference: 3685824
MDL No.: MFCD00038998
Formula:
C₄H₉NO₂S·HCl Melting Pt: 142 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038998 CAS Number: 18598-63-5 EINECS: 242-435-0 Merck Index: 14,5785 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...