Locostatin

Supplier: Thermo Scientific Chemicals

J64425-MA J64425-MF
CAAAJ64425-MAEA 222.78 CAD
CAAAJ64425-MA CAAAJ64425-MF
Locostatin
Locostatin

Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration


Powder


Applications: Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration

Formula: C₁₄H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00278769
CAS Number: 90719-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR