Ammonium sulfate 99+%, Ultrapure
Supplier: Thermo Scientific Chemicals
J64419-22
J64419-36
J64419-A3
CAAAJ64419-22EA
51.19
CAD
CAAAJ64419-22
CAAAJ64419-36
CAAAJ64419-A3
Ammonium sulfate 99+%, Ultrapure
Ammonium sulphate
Crystalline
Purity: 99+%
CAS No.: 7783-20-2
Molecular Formula: H8N2O4S
Formula Weight: 132.14
Physical Form: Crystalline
Density: 1.769
Appearance: White
Melting Point: ca 280° dec.
Merck Reference: 14,555
MDL No.: MFCD00003391
Formula:
(NH₄)₂SO₄ Melting Pt: ∼ 280 °C (dec.) Density: 1.769 Storage Temperature: Ambient |
MDL Number:
MFCD00003391 CAS Number: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 14,555 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...