2'-Deoxyadenosine-5'-monophosphate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
dAMP, ((2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate
J64402-03
J64402-06
CAAAJ64402-03EA
152.31
CAD
CAAAJ64402-03
CAAAJ64402-06
2'-Deoxyadenosine-5'-monophosphate 98%
2'-Deoxyadenosine-5'-monophosphate
Crystalline powder
Purity: 98%
CAS No.: 653-63-4
Molecular Formula: C10H14N5O6P
Formula Weight: 331.22
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD00005753
Formula:
C₁₀H₁₄N₅O₆P MW: 331.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005753 CAS Number: 653-63-4 EINECS: 211-503-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...