Uridine-5'-diphosphate disodium salt 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64329-03
J64329-MC
CAAAJ64329-03EA
183.28
CAD
CAAAJ64329-03
CAAAJ64329-MC
Uridine-5'-diphosphate disodium salt 98%
Uridine-5'-diphosphate disodium salt
Powder
Purity: 98%
CAS No.: 27821-45-0
Molecular Formula: C9H12N2Na2O12P2
Formula Weight: 448.13
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to off-white
Beilstein Reference: 8817400
MDL No.: MFCD00149253
Formula:
C₉H₁₂N₂Na₂O₁₂P₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00149253 CAS Number: 27821-45-0 EINECS: 248-678-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...